Skip to Content
Merck
All Photos(2)

Key Documents

GF60507019

Lanthanum

rod, 50mm, diameter 6.35mm, cast, 99%

Synonym(s):

Lanthanum, LA007910

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
La
CAS Number:
Molecular Weight:
138.91
MDL number:
UNSPSC Code:
12141607
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

rod

manufacturer/tradename

Goodfellow 605-070-19

resistivity

54 μΩ-cm

L × diam.

50 mm × 6.35 mm

bp

3464 °C (lit.)

mp

920 °C (lit.)

density

6.19 g/mL at 25 °C (lit.)

SMILES string

[La]

InChI

1S/La

InChI key

FZLIPJUXYLNCLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stuart M Sprague
Current medical research and opinion, 23(12), 3167-3175 (2007-11-10)
Obstacles to successful management of hyperphosphatemia in chronic kidney disease include inadequate control of dietary phosphate and non-compliance with phosphate-binder therapy. Three major classes of phosphate binders include calcium-based binders, sevelamer HCl, and lanthanum carbonate. A literature search was performed
Tilman B Drüeke
Seminars in dialysis, 20(4), 329-332 (2007-07-20)
Controlling serum phosphorus levels in patients with renal failure is critical. The use of oral phosphate-binding agents is universal for patients with end-stage kidney disease to reduce phosphate absorption. The therapeutic goal is to reduce serum phosphorus levels without disturbing
Amy Barton Pai et al.
Expert opinion on drug metabolism & toxicology, 5(1), 71-81 (2009-02-25)
Hyperphosphatemia is recognized as a principal mineral disorder in chronic kidney disease (CKD) that leads to the development of secondary hyperparathyroidism. Recent data indicate that hyperphosphatemia is associated with accelerated cardiac calcification and increased mortality in patients with CKD. Control
Gadolinium and lanthanum: a iatrogenic transmetallation?
Simona Brambilla et al.
Clinical biochemistry, 41(13), 1029-1033 (2008-06-19)
M Cozzolino et al.
The International journal of artificial organs, 30(4), 293-300 (2007-05-24)
Phosphate overload is a dramatic consequence in end-stage renal disease (ESRD) patients. Recent studies have well documented that abnormalities in mineral and bone metabolism in these patients are associated with increased cardiovascular morbidity and mortality. Elevated serum phosphate and calcium-phosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service