Skip to Content
Merck
All Photos(1)

Key Documents

GF48659844

Copper-zirconium alloy, Cu70Zr30

powder, 150micron max. particle size, alloy pre-cursor, 100g

Synonym(s):

CU236010

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
31152304
NACRES:
NA.23

form

powder

manufacturer/tradename

Goodfellow 486-598-44

weight

100 g

max. part. size

150 μm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service