GF48047722
Tungsten carbide
powder, max. particle size 45 micron, weight 50 g, purity 99.5%
Synonym(s):
W 526011
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
31191518
NACRES:
NA.23
Recommended Products
Assay
99.5%
form
powder
manufacturer/tradename
Goodfellow 480-477-22
weight
50 g
max. part. size
45 μm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service