GF23000498
Monel® alloy k-500, Ni63Cu30Al3Fe2Mn1.5Ti0.5
sheet, 200 x 200mm, 3.5mm thickness, annealed
Synonym(s):
NE043050
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101712
NACRES:
NA.23
Recommended Products
form
foil
manufacturer/tradename
Goodfellow 230-004-98
L × W × thickness
200 mm × 200 mm × 3.5 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Monel is a registered trademark of Special Metals Corporation
Monel is a registered trademark of Huntington Alloys Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service