GF19020620
Tin
rod, 500mm, diameter 3.0mm, 99.99+%
Synonym(s):
Tin, SN007918
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.99%
form
rod
manufacturer/tradename
Goodfellow 190-206-20
resistivity
11 μΩ-cm, 20°C
L × diam.
500 mm × 3.0 mm
bp
2270 °C (lit.)
mp
231.9 °C (lit.)
density
7.310 g/mL at 25 °C (lit.)
SMILES string
[Sn]
InChI
1S/Sn
InChI key
ATJFFYVFTNAWJD-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Applications of tin-containing intermediates to carbohydrate chemistry.
T B Grindley
Advances in carbohydrate chemistry and biochemistry, 53, 17-142 (1998-08-26)
Y Arakawa
Sangyo eiseigaku zasshi = Journal of occupational health, 39(1), 1-20 (1997-01-01)
Tin generates a wide variety of biological activities deriving from its chemical character. In this article, the biological activities of tin compounds are reviewed with a focus on the connection with immunity. The table of contents is as follows: Introduction
The newer essential trace elements, chromium, tin, nickel, vanadium and silicon.
W Mertz
The Proceedings of the Nutrition Society, 33(3), 307-313 (1974-12-01)
[Tin in the environment].
Iu P Popov et al.
Gigiena i sanitariia, (9)(9), 55-57 (1983-09-01)
K A Winship
Adverse drug reactions and acute poisoning reviews, 7(1), 19-38 (1988-01-01)
Inorganic tin salts are poorly absorbed and rapidly excreted in the faeces; as a result they have a low toxicity. Only about 5 per cent is absorbed from the gastrointestinal tract, widely distributed in the body, then excreted by the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service