Skip to Content
Merck
All Photos(2)

Key Documents

GF13609111

Lanthanum

foil, 25x25mm, thickness 0.125mm, as rolled, 99%

Synonym(s):

Lanthanum, LA000215

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
La
CAS Number:
Molecular Weight:
138.91
MDL number:
UNSPSC Code:
12141607
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

foil

manufacturer/tradename

Goodfellow 136-091-11

resistivity

54 μΩ-cm

size × thickness

25 x 25 mm × 0.125 mm

bp

3464 °C (lit.)

mp

920 °C (lit.)

density

6.19 g/mL at 25 °C (lit.)

SMILES string

[La]

InChI

1S/La

InChI key

FZLIPJUXYLNCLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stuart M Sprague
Current medical research and opinion, 23(12), 3167-3175 (2007-11-10)
Obstacles to successful management of hyperphosphatemia in chronic kidney disease include inadequate control of dietary phosphate and non-compliance with phosphate-binder therapy. Three major classes of phosphate binders include calcium-based binders, sevelamer HCl, and lanthanum carbonate. A literature search was performed
Yongsheng Yang et al.
Molecular pharmaceutics, 10(2), 544-550 (2012-12-20)
A conventional human pharmacokinetic (PK) in vivo study is often considered as the "gold standard" to determine bioequivalence (BE) of drug products. However, this BE approach is not always applicable to the products not intended to be delivered into the
Monique P Curran et al.
Drugs, 69(16), 2329-2349 (2009-10-27)
Orally administered lanthanum carbonate (Fosrenol) dissociates in the acid environment of the upper gastrointestinal tract to release the cation lanthanum, which then binds dietary phosphate. Lanthanum carbonate was effective in reducing levels of serum phosphate and serum calcium x phosphate
Marc E De Broe
Seminars in dialysis, 21(2), 142-144 (2008-02-06)
The recent recognition of gadolinium (Gd) administration as a cause of nephrogenic systemic fibrosis (NSF) is a growing concern. The similarity of Gd and lanthanum (La), based on their place in the table of Mendeljev, has led to assumptions that
Tilman B Drüeke
Seminars in dialysis, 20(4), 329-332 (2007-07-20)
Controlling serum phosphorus levels in patients with renal failure is critical. The use of oral phosphate-binding agents is universal for patients with end-stage kidney disease to reduce phosphate absorption. The therapeutic goal is to reduce serum phosphorus levels without disturbing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service