Skip to Content
Merck
All Photos(1)

Key Documents

F20408

Sigma-Aldrich

2-Furanmethanethiol

98%

Synonym(s):

2-Furfurylthiol, 2-Furylmethanethiol, Furfuryl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
Beilstein:
383594
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.531 (lit.)

bp

155 °C (lit.)

density

1.132 g/mL at 25 °C (lit.)

SMILES string

SCc1ccco1

InChI

1S/C5H6OS/c7-4-5-2-1-3-6-5/h1-3,7H,4H2

InChI key

ZFFTZDQKIXPDAF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imre Blank et al.
Journal of agricultural and food chemistry, 50(8), 2356-2364 (2002-04-04)
The stability of the coffee flavor compound furfuryl mercaptan has been investigated in aqueous solutions under Fenton-type reaction conditions. The impact of hydrogen peroxide, iron, ascorbic acid, and ethylenediaminetetraacetic acid was studied in various combinations of reagents and temperature. Furfuryl
Brian G Lake et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(12), 1761-1770 (2003-10-18)
The metabolism of two thiofurans, namely furfuryl mercaptan (FM) and 2-methyl-3-furanthiol (MTF), to their corresponding methyl sulphide and methyl sulphoxide derivatives has been studied in male Sprague-Dawley rat hepatocytes and liver microsomes. Rat hepatocytes converted FM to furfuryl methyl sulphoxide
Christoph Müller et al.
Journal of agricultural and food chemistry, 53(7), 2623-2629 (2005-03-31)
The purpose of the following study was to investigate the influence of coffee roasting on the thiol-binding activity of coffee beverages, and to investigate the potential of various green bean compounds as precursors of thiol-binding sites by using promising "in
Christoph Müller et al.
Journal of agricultural and food chemistry, 55(10), 4095-4102 (2007-04-19)
To gain a more comprehensive knowledge of the contribution of recently identified phenol/thiol conjugates to the storage-induced degradation of odorous thiols, the concentrations of the sulfury-roasty smelling key odorant 2-furfurylthiol and the concentrations of the putative thiol-receptive di- and trihydroxybenzenes
Luigi Poisson et al.
Journal of agricultural and food chemistry, 57(21), 9923-9931 (2009-10-13)
The formation of several key odorants, such as 2-furfurylthiol (FFT), alkylpyrazines, and diketones, was studied upon coffee roasting. The approach involved the incorporation of potential precursors in green coffee beans by means of biomimetic in-bean and spiking experiments. Both labeled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service