These electrodes are grade 304 stainless steel. Grade 304 stainless steel is generally regarded as the most common austenitic stainless steel. This grade contains a high nickel content, typically between 8 and 10.5 percent by weight, and a high amount of chromium, approximately 18 to 20 percent by weight. The high amounts of chromium and nickel give 304 stainless steel excellent corrosion resistance.
ESYNTH010
SynLectro™ Stainless Steel Electrode
size 3 mm × 20 mm × 60 mm
About This Item
Recommended Products
size
3 mm × 20 mm × 60 mm
General description
SynLectro™ Electrolysis Platform
The SynLectro™ Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Related Content
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
-
What is the grade of your SynLectro Stainless Steel Electrodes?
1 answer-
Helpful?
-