Skip to Content
Merck
All Photos(1)

Key Documents

DWKBLH0100P

Azlon HDPE Wide Neck Round Bottles

screw cap, capacity 100 mL

Synonym(s):

BLH0100P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121818
NACRES:
NB.43

material

screw cap
wide-neck

packaging

pkg of 10 ea

manufacturer/tradename

Azlon BLH0100P

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for management and storage of both liquid and powder samples

Features and Benefits

Suitable for food contact applications

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service