D8801
2,6-Diacetylpyridine
98%
Synonym(s):
1,1′-(2,6-Pyridinediyl)diethanone, 1-(6-Acetylpyridin-2-yl)ethanone
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17
Beilstein:
118286
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
79-82 °C (lit.)
SMILES string
CC(=O)c1cccc(n1)C(C)=O
InChI
1S/C9H9NO2/c1-6(11)8-4-3-5-9(10-8)7(2)12/h3-5H,1-2H3
InChI key
BEZVGIHGZPLGBL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
2,6-Diacetylpyridine can be used as a starting material for the synthesis of:
- Iron and cobalt bis(imino)pyridyl complexes, catalysts for ethylene oligomerization.
- Thiosemicarbazone derivatives with potential application as anti-mycobacterial tuberculosis agent.
- Macrocyclic pentadentate nitrogen–sulphur donor ligands by [1+1] condensation with 1,2-di(o-aminophenylthio)ethane.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Oligomerisation of Ethylene by Bis (imino) pyridyliron and-cobalt Complexes.
Britovsek GJP, et al.
Chemistry?A European Journal , 6(12), 2221-2231 (2000)
Thiosemicarbazones, semicarbazones, dithiocarbazates and hydrazide/hydrazones: Anti-Mycobacterium tuberculosis activity and cytotoxicity.
Pavan FR, et al.
European Journal of Medicinal Chemistry, 45(5), 1898-1905 (2010)
Manja Kubeil et al.
ChemistryOpen, 8(5), 637-642 (2019-05-30)
A photoactivatable ruthenium(II) carbonyl complex mer,cis-[Ru(II)Cl(BisQ)(CO)2]PF62 was prepared using a tridentate bisquinoline ligand (BisQ=(2,6-diquinolin-2-yl)pyridin). Compound 2 was thoroughly characterized by standard analytical methods and single crystal X-ray diffraction. The crystal structure of the complex cation reveals a distorted octahedral geometry.
Chromium (III), manganese (II), iron (III), cobalt (II), nickel (II) and copper (II) complexes with a pentadentate, 15-membered new macrocyclic ligand.
Chandra S and Gupta K
Transition Metal Chemistry, 27(2), 196-199 (2002)
Ali Sayqal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 242, 118711-118711 (2020-08-02)
Herein, the presented manuscript provides for an extensive spectrofluorimetric method for micro determination of silver ion. This established method based on the use of the three synthesized 2,6-disubstituted pyridine derivatives (R1, R2 and R3) through exploiting their high fluorescence emission
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service