Skip to Content
Merck
All Photos(5)

Key Documents

D7881

Sigma-Aldrich

meso-2,3-Dimercaptosuccinic acid

~98%

Synonym(s):

2,3-Dithio-meso-tartaric acid, DIM-SA, DMS, DMSA, Succimer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(SH)CH(SH)CO2H
CAS Number:
Molecular Weight:
182.22
Beilstein:
1725150
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

~98%

form

powder

mp

196-198 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

OC(=O)[C@@H](S)[C@@H](S)C(O)=O

InChI

1S/C4H6O4S2/c5-3(6)1(9)2(10)4(7)8/h1-2,9-10H,(H,5,6)(H,7,8)/t1-,2+

InChI key

ACTRVOBWPAIOHC-XIXRPRMCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

meso-2,3-Dimercaptosuccinic acid (DMSA) has been used to form a monolayer on Mg/Ti/Au micromotors to enhance the chelation of toxic heavy metal contaminants (Zn(II), Cd(II) and Pb(II)) from polluted aqueous solutions.
It can be used:
  • As a reducing and stabilizing agent to synthesize monolayer-protected gold clusters.
  • For terminal carboxylation of the oleylamine coated Fe3O4 nanoparticles, to develop targeted curcumin drug delivery system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-pot preparation of subnanometer-sized gold clusters via reduction and stabilization by meso-2, 3-dimercaptosuccinic acid.
Negishi Y and Tsukuda T
Journal of the American Chemical Society, 125(14), 4046-4047 (2003)
Superparamagnetic Fe3O4 nanoparticles: synthesis by a solvothermal process and functionalization for a magnetic targeted curcumin delivery system.
Qi M, et al.
New. J. Chem., 40(5), 4480-4491 (2016)
Thanasis Chouliaras et al.
Membranes, 9(12) (2019-12-11)
Polymeric (ionic liquid) (PIL) copolymers bearing cationic imidazolium pendants and polar acrylic acid groups (P(VBCImY-co-AAx)), which both favor the interaction with CO2 molecules, have been synthesized and blended with film forming, high glass transition temperature aromatic polyether-based pyridinium PILs (PILPyr).
Francesca Brero et al.
Nanomaterials (Basel, Switzerland), 10(10) (2020-10-01)
A combination of carbon ions/photons irradiation and hyperthermia as a novel therapeutic approach for the in-vitro treatment of pancreatic cancer BxPC3 cells is presented. The radiation doses used are 0-2 Gy for carbon ions and 0-7 Gy for 6 MV
Self-propelled chelation platforms for efficient removal of toxic metals.
Uygun DA, et al.
Environmental science. Nano, 3(3), 559-566 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service