D21405
1,8-Diaminonaphthalene
99%
Synonym(s):
1,8-Naphthalenediamine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C10H6(NH2)2
CAS Number:
Molecular Weight:
158.20
Beilstein:
742841
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
crystals
bp
205 °C/12 mmHg (lit.)
mp
60-65 °C (lit.)
SMILES string
Nc1cccc2cccc(N)c12
InChI
1S/C10H10N2/c11-8-5-1-3-7-4-2-6-9(12)10(7)8/h1-6H,11-12H2
InChI key
YFOOEYJGMMJJLS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
>392.0 °F - closed cup
Flash Point(C)
> 200 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Guanzhao Wu et al.
Research (Washington, D.C.), 2019, 6717104-6717104 (2019-09-25)
3D chirality of sandwich type of organic molecules has been discovered. The key element of this chirality is characterized by three layers of structures that are arranged nearly in parallel fashion with one on top and one down from the
Kai Schickedanz et al.
Chemical communications (Cambridge, England), 51(87), 15808-15810 (2015-09-15)
The high-yield synthesis of a boron-doped [4]helicene was achieved through a Ni-mediated Yamamoto C-C-coupling reaction; the moderate distortion of the molecular scaffold confers excellent solubility to the air- and water-stable green luminophore.
Liliana Martínez-Ávila et al.
Environmental pollution (Barking, Essex : 1987), 271, 116358-116358 (2021-01-02)
Polyaromatic phenanthrene (Phe) and benzo[a]pyrene (BaP) are highly toxic, mutagenic, and carcinogenic contaminants widely dispersed in nature, including saline environments. Polyextremotolerant Rhodotorula mucilaginosa EXF-1630, isolated from Arctic sea ice, was grown on a huge concentration range -10 to 500 ppm- of
Roman Papoušek et al.
Journal of separation science, 42(8), 1629-1633 (2019-02-15)
Masking derivatization was introduced for the determination of residual solvents in samples containing a volatile reactive matrix component(s). Isobutylboronic acid, used in the last step of Bortezomib synthesis, represents a compound passing to the gas phase and deteriorating a chromatographic
Mohan N Patel et al.
Bioorganic & medicinal chemistry, 17(15), 5648-5655 (2009-07-08)
Here in we tried to increase an antibacterial activity of ciprofloxacin drug due to formation of mixed-ligand complexes. Synthesized compounds were found to be more potent compare to drugs, ligands and metal salt against selective gram((+ve)) and gram((-ve)) organisms. Interaction
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service