Skip to Content
Merck
All Photos(4)

Key Documents

D175404

Sigma-Aldrich

3,4-Dimethylphenol

98%

Synonym(s):

1,2-Dimethyl-4-hydroxybenzene, 3,4-Xylenol, 4-Hydroxy-o-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1099267
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

crystals

bp

227 °C (lit.)

mp

65-68 °C

storage temp.

2-8°C

SMILES string

Cc1ccc(O)cc1C

InChI

1S/C8H10O/c1-6-3-4-8(9)5-7(6)2/h3-5,9H,1-2H3

InChI key

YCOXTKKNXUZSKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Applications of 3,4-dimethylphenol:
  • It can react with ethyl cinnamates in trifluoroacetic acid to form the corresponding dihydrocoumarin derivatives.[1]
  • Bromination of 3,4-dimethylphenol using bromine can lead to 6-bromo-3,4-dimethylphenol.[2]
  • It can react with 1-fluoro-4-chloromethyl-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoro-borate) (Selectfluor® F-TEDA-BF4) to form 4-fluoro-3,4-dimethylcyclohexa-2,5-dienone.[3]

Legal Information

Selectfluor is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Synthesis of 4-Fluorocyclohexa-2, 5-dienone Derivatives using N-fluoro-1, 4-diazoniabicyclo [2.2. 2] octane Salt Analogues
Stavber S, et al.
Synlett, 1999(09), 1375-1378 (1999)
A convenient synthesis of dihydrocoumarins from phenols and cinnamic acid derivatives
Aoki S, et al.
Tetrahedron, 61(39), 9291-9297 (2005)
Synthesis, x-ray structures, conformational fixation, and proton NMR spectroscopic studies.
Andreetti G D, et al.
The Journal of Organic Chemistry, 58(15), 4023-4032 (1993)
M Bartilson et al.
Molecular & general genetics : MGG, 220(2), 294-300 (1990-01-01)
The gene organization of the phenol catabolic pathway of Pseudomonas CF600 has been investigated. This strain can grow on phenol and some methylated phenols by virtue of an inducible phenol hydroxylase and metacleavage pathway enzymes. The genes coding for these
A P Hart et al.
Journal of forensic sciences, 42(4), 693-696 (1997-07-01)
Phenol (carbolic acid) is widely used as a disinfectant as well as in the chemical industry as an intermediate in the synthesis of a variety of chemicals. Phenol is also the major metabolite of benzene which is used in many

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service