Skip to Content
Merck
All Photos(1)

Documents

D170305

Sigma-Aldrich

1,4-Dimethylnaphthalene

95%

Synonym(s):

1,4-Dimethylnaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CH3)2
CAS Number:
Molecular Weight:
156.22
Beilstein:
2039842
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.613 (lit.)

bp

262-264 °C/751 mmHg (lit.)

mp

−18 °C (lit.)

density

1.016 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)c2ccccc12

InChI

1S/C12H12/c1-9-7-8-10(2)12-6-4-3-5-11(9)12/h3-8H,1-2H3

InChI key

APQSQLNWAIULLK-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

D S Malament et al.
Carcinogenesis, 2(8), 723-729 (1981-01-01)
Phenanthrene (Phe) and to a lesser degree 1,4-dimethylnaphthalene (DMeN) were each found to retard the development of epidermoid carcinomas in hamster buccal pouch induced by the thrice weekly application of a 0.5 per cent solution of 7,12-dimethylbenz[a]anthracene (DMBA) in heavy
Michael A Campbell et al.
Functional & integrative genomics, 12(3), 533-541 (2011-11-25)
The suppression of sprout growth is critical for the long-term storage of potato tubers. 1,4-Dimethylenapthlene (DMN) is a new class of sprout control agent but the metabolic mode of action for this compound has yet to be elucidated. Changes in
Michael A Campbell et al.
Plant molecular biology, 73(1-2), 181-189 (2010-02-06)
Chlorpropham (CIPC) and 1,4-dimethylnapthalene (DMN) are used to control postharvest sprouting of potato tubers. CIPC alters microtubule structure and function resulting in inhibition of cell division. The mechanism of action of DMN is unknown but, because it is a natural
Damir Posavec et al.
Organic & biomolecular chemistry, 10(35), 7062-7069 (2012-08-01)
Decomposition of endoperoxide containing molecules is an attractive approach for the delayed release of singlet oxygen under mild reaction conditions. Here we describe a new method for the adaptation of the corresponding decay times by controlling the supramolecular functional structure
P Di Mascio et al.
Biological chemistry Hoppe-Seyler, 376(5), 297-301 (1995-05-01)
Carotenoids comprise one of the most widespread classes of pigments found in nature. Polyene pigments from the dinoflagellate Gonyaulax polyedra were extracted every hour over a 24 hour period and the levels of beta-carotene during the day-phase were found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service