Skip to Content
Merck
All Photos(6)

Key Documents

D157805

Sigma-Aldrich

N,N′-Dimethylethylenediamine

98%

Synonym(s):

DMEDA, 1,2-Bis(methylamino)ethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCH2CH2NHCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
878142
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.431 (lit.)

bp

119 °C (lit.)

density

0.819 g/mL at 20 °C (lit.)

SMILES string

CNCCNC

InChI

1S/C4H12N2/c1-5-3-4-6-2/h5-6H,3-4H2,1-2H3

InChI key

KVKFRMCSXWQSNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

N,N′-Dimethylethylenediamine can be used as a ligand in the following processes:
  • Copper-catalyzed cyanations of aryl bromides using potassium hexacyanoferrate(II) to form the corresponding benzonitriles.
  • Copper-catalyzed tandem reaction of carbodiimide and isocyanoacetate to form benzoimidazo[1,5-α]imidazoles.
  • Aminothiolation of 1,1-dibromoalkene to form N-fused heterocycles.
  • In combination with ferrous chloride, for the N-arylation of NH-sulfoximines with aryl iodides to form N-arylsulfoximines.
  • It can catalyze the direct C-H arylation of unactivated benzene in the presence of potassium tert-butoxide to form the corresponding biaryl.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organocatalysis in cross-coupling: DMEDA-catalyzed direct C? H arylation of unactivated benzene.
Liu W, et al.
Journal of the American Chemical Society, 132(47), 16737-16740 (2010)
Ghotas Evindar et al.
The Journal of organic chemistry, 71(5), 1802-1808 (2006-02-25)
A general method for the formation of benzoxazoles via a copper-catalyzed cyclization of ortho-haloanilides is reported. This approach complements the more commonly used strategies for benzoxazole formation which require 2-aminophenols as substrates. The reaction involves an intramolecular C-O cross-coupling of
Kim A Lennox et al.
Oligonucleotides, 16(1), 26-42 (2006-04-06)
A wide variety of modified oligonucleotides have been tested as antisense agents. Each chemical modification produces a distinct profile of potency, toxicity, and specificity. Novel cationic phosphoramidate-modified antisense oligonucleotides have been developed recently that have unique and interesting properties. We
Iron?Catalyzed C N Cross?Coupling of Sulfoximines with Aryl Iodides.
Correa A, et al.
Advanced Synthesis & Catalysis, 350(3), 391-394 (2008)
An environmentally benign procedure for the Cu-catalyzed cyanation of aryl bromides.
Schareina T, et al.
Tetrahedron Letters, 46(15), 2585-2588 (2005)

Articles

Buchwald phosphine ligands for C-C, C-N, and C-O bond formation.

Buchwald phosphine ligands for C-C, C-N, and C-O bond formation.

Buchwald phosphine ligands for C-C, C-N, and C-O bond formation.

Buchwald phosphine ligands for C-C, C-N, and C-O bond formation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service