D134007
3,5-Dimethoxycinnamic acid, predominantly trans
≥99%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3O)2C6H3CH=CHCO2H
CAS Number:
Molecular Weight:
208.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥99%
form
solid
mp
174-175 °C (lit.)
SMILES string
COc1cc(OC)cc(\C=C\C(O)=O)c1
InChI
1S/C11H12O4/c1-14-9-5-8(3-4-11(12)13)6-10(7-9)15-2/h3-7H,1-2H3,(H,12,13)/b4-3+
InChI key
VLSRUFWCGBMYDJ-ONEGZZNKSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Louise C Simms et al.
Pest management science, 58(7), 687-694 (2002-07-31)
Slugs are major pests of oilseed rape that are poorly controlled by conventional bait pellets. A series of laboratory experiments investigated the potential of seed-dressings to control slug damage in this crop. Four compounds: metaldehyde, methiocarb, cinnamamide and 3,5-dimethoxycinnamic acid
T W Rosanske et al.
Journal of pharmaceutical sciences, 69(5), 564-567 (1980-05-01)
The solubility, spectral, and kinetic methods were used to study complexing between alpha-cyclodextrin (ligand, L) and 3,5-dimethoxycinnamic acid, benzalacetone, and methyl cinnamate (substrates, S). In aqueous solution at 25 degrees and with an ionic strength of 0.01 M, the following
Katsunori Teranishi
Luminescence : the journal of biological and chemical luminescence, 32(2), 182-189 (2016-06-09)
Mycena chlorophos is an oxygen-dependent bioluminescent fungus. The mechanisms underlying its light emission are unknown. A component that increased the bioluminescence intensity of the immature living gills of M. chlorophos was isolated from mature M. chlorophos gills and chemically characterized.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service