Skip to Content
Merck
All Photos(5)

Documents

D1133

Sigma-Aldrich

Diethylenetriaminepentaacetic acid

≥98% (titration)

Synonym(s):

(Carboxymethylimino)bis(ethylenenitrilo)tetraacetic acid, N,N-Bis(2-[bis(carboxymethyl)amino]ethyl)glycine, DETAPAC, DTPA, Penta(carboxymethyl)diethylenetriamine, Pentetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(HOOCCH2)2NCH2CH2]2NCH2COOH
CAS Number:
Molecular Weight:
393.35
Beilstein:
1810219
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic (organic)

Quality Level

Assay

≥98% (titration)

form

powder

mp

219-220 °C (lit.)

solubility

1 M HCl: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O

InChI

1S/C14H23N3O10/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27)

InChI key

QPCDCPDFJACHGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylenetriaminepentaacetic acid may be used to synthesize chitosan (CH)-gadolinium (Gd) diethylenetriaminepentaacetic acid (DTPA) conjugates.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

200 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P C Adams et al.
Hepatology (Baltimore, Md.), 14(6), 1230-1234 (1991-12-01)
The elimination of toxic metal ions metabolically accumulated by patients remains a difficult clinical problem and a target of drug development. DTPA (diethylenetriaminepentaacetic acid) is a hydrophilic chelating agent with high affinity for divalent and trivalent metal ions including iron
Gauthier J-P Deblonde et al.
Nature communications, 10(1), 2438-2438 (2019-06-06)
Metal ion separations are critical to numerous fields, including nuclear medicine, waste recycling, space exploration, and fundamental research. Nonetheless, operational conditions and performance are limited, imposing compromises between recovery, purity, and cost. Siderophore-inspired ligands show unprecedented charge-based selectivity and compatibility
Maria P Sciaudone et al.
Biological trace element research, 99(1-3), 219-231 (2004-07-06)
Zinc is thought to be required as a structural component of the thyroid hormone (triiodothyronine, T3) receptor (TR). However, we have previously demonstrated that use of diethylenetriaminepentaacetic acid (DTPA) to restrict zinc availability to cultured cells actually potentiates rather than
Kosmas Vamvakidis et al.
Materials (Basel, Switzerland), 13(7) (2020-04-02)
Water pollution by heavy metals is one of the most serious worldwide environmental issues. With a focus on copper(II) ions and copper complex removal, in the present study, ultra-small primary CoFe2O4 magnetic nanoparticles (MNPs) coated with octadecylamine (ODA) of adequate
Chitosan modified with gadolinium diethylenetriaminepentaacetic acid for magnetic resonance imaging of DNA/chitosan nanoparticles.
Darras V, et al.
Carbohydrate Polymers, 80(4), 1137-1146 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service