Skip to Content
Merck
All Photos(3)

Key Documents

C69802

Sigma-Aldrich

2-Chloropyridine

99%

Synonym(s):

α-Chloropyridine, o-Chloropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4ClN
CAS Number:
Molecular Weight:
113.54
Beilstein:
105788
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1.7 mmHg ( 20 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.532 (lit.)

bp

166 °C/714 mmHg (lit.)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccn1

InChI

1S/C5H4ClN/c6-5-3-1-2-4-7-5/h1-4H

InChI key

OKDGRDCXVWSXDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fereshteh Mehri et al.
Iranian journal of pharmaceutical research : IJPR, 19(4), 143-150 (2021-04-13)
A sensitive method using ion-pair extraction was developed by liquid chromatography tandem mass spectrometry (LC-MS/MS) for measurement of 4-methylimidazole (4-MI) in NMRI mice plasma and cerebrospinal fluid (CSF). Detection was done by electrospray positive ionization mass spectrometry in the multiple-reaction
Kazuko Inoue et al.
Drug metabolism and disposition: the biological fate of chemicals, 37(9), 1797-1800 (2009-06-23)
We investigated the substitution reaction of a series of 2-chloropyridine derivatives catalyzed by rat liver microsomal glutathione S-transferase 1. Various 2-chloropyridine derivatives were metabolized to the corresponding substituted glutathione conjugates via displacement of chlorine atom with glutathione. The reaction was
David R Stapleton et al.
Water research, 43(16), 3964-3973 (2009-07-14)
The photolytic destruction and mineralisation of 2-halogenated pyridines (2-HalPYs) was studied in a range of conditions by means of ultraviolet irradiation at 254 nm. In all cases pseudo-first order kinetics satisfactorily describe the degradation process. Key decomposition products were identified
Andreas Reichelt et al.
Organic letters, 12(4), 792-795 (2010-01-27)
An efficient and convenient method for the synthesis of [1,2,4]triazolo[4,3-a]pyridines was exemplified by the synthesis of 20 analogues bearing a variety of substituents at the 3-position. The methodology involves a palladium-catalyzed addition of hydrazides to 2-chloropyridine, which occurs chemoselectively at
Contribution of N-oxidation and .OH radicals to mutagenesis of 2-chloropyridine in Salmonella typhimurium.
B Chłopkiewicz et al.
Acta biochimica Polonica, 40(1), 57-59 (1993-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service