C105058
Chlorocyclohexane
99%
Synonym(s):
Cyclohexyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H11Cl
CAS Number:
Molecular Weight:
118.60
Beilstein:
1900796
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
99%
form
liquid
refractive index
n20/D 1.462 (lit.)
bp
142 °C (lit.)
mp
−44 °C (lit.)
density
1 g/mL at 25 °C (lit.)
SMILES string
ClC1CCCCC1
InChI
1S/C6H11Cl/c7-6-4-2-1-3-5-6/h6H,1-5H2
InChI key
UNFUYWDGSFDHCW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
93.2 °F
Flash Point(C)
34 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Henriëtte E Bakker et al.
Langmuir : the ACS journal of surfaces and colloids, 33(4), 881-890 (2017-01-04)
The electrophoretic mobility and the zeta potential (ζ) of fluorescently labeled colloidal silica rods, with an aspect ratio of 3.8 and 6.1, were determined with microelectrophoresis measurements using confocal microscopy. In the case where the colloidal particles all move at
Octavio Perez-Garcia et al.
Environmental science & technology, 51(23), 13678-13688 (2017-10-31)
Metabolomics provides insights into the actual physiology of cells rather than their mere "potential", as provided by genomic and transcriptomic analysis. We investigate the modulation of nitrous oxide (N
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service