BR82220
BRAND® needle-valve stopcocks for burettes and automatic burettes with PTFE spindle
Straight, for burette capacity 2-10 mL
Synonym(s):
Accessories, Burettes, Needle-valve stopcock, Stopcocks
All Photos(4)
About This Item
UNSPSC Code:
40141602
NACRES:
NB.43
Recommended Products
material
polystyrene
feature
for burette capacity 2-10 mL
packaging
pack of 1 ea
manufacturer/tradename
BRAND 82220
General description
Interchangeable PTFE valve spindle, PP cap and retention device. Valve opening 0-25.5 mm.
Please note: PTFE valve spindle operates smoothly yet is tight, even without lubrication, so there is no grease residue on the burette. The valve spindle is replaceable. Tight seal without force, a gentle turn is all it takes to close the burette valve.
Please note: PTFE valve spindle operates smoothly yet is tight, even without lubrication, so there is no grease residue on the burette. The valve spindle is replaceable. Tight seal without force, a gentle turn is all it takes to close the burette valve.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)