BR781701
BRAND® microplate BRANDplates®, pureGrade
size 1536 wells, size 10 μL, flat bottom, white
Synonym(s):
BRANDplates®, Microplates, pureGradeS
About This Item
polystyrene
white wells
1536 wells
Recommended Products
material
flat bottom
polystyrene
white wells
sterility
sterile
feature
lid
skirt (F-bottom)
packaging
pack of 50 ea
pkg of (individually packaged)
manufacturer/tradename
BRAND 781701
size
10 μL
1536 wells
working volume
10 μL
color
white
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service