BR701340
BRAND® 96-well deep well plate, stackable
volume 1.2 mL, polypropylene, low profile
Synonym(s):
Deep-well Plates, Sample Storage
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
polypropylene
round bottom wells
round bottom wells
size:
96 well
volume:
1.2 mL
working volume:
1.2 mL
sterility:
non-sterile
binding type:
non-treated surface
feature:
skirt
Recommended Products
material
polypropylene
round bottom wells
description
alphanumeric identification system
sterility
non-sterile
feature
skirt
packaging
pack of 50 ea
manufacturer/tradename
BRAND 701340
size
96 well
volume
1.2 mL
working volume
1.2 mL
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- BRAND® 96-well deep well plates are made of high-grade chemically resistant polypropylene.
- Ideal for use in various automated processes including High-Throughput Screening (HTS), cell and tissue cultures, serial dilutions, reagent transfer, and sample storage down to -80 °C.
- The raised edges of the well facilitate a secure seal and prevent sample contamination.
- Stackable - A convenient space-saving option.
- ′U′ bottom base ensures optimal sample mixing and collection.
- Alphanumeric coding for easy identification.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service