Key Documents
BR134080
BRAND® flow indicator
for tubing I.D. 6-11 mm
Synonym(s):
Flow indicator, Tubing
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
material
polypropylene
packaging
pack of 1 ea
manufacturer/tradename
BRAND 134080
Related Categories
General description
Styrene- acrylonitrile (SAN), transparent. This useful device optimizes cooling water flow during distillation. This helps to decrease operating costs by reducing water consumption. Operating pressure max. 2 bar, operating temperature for SAN max. 30 °C. For tubing of inner-Ø 6-11 mm.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)