BAF371250078
Spinfin® magnetic stirring bar
H × diam. 0.5 in. (12.7 mm) × 0.875 in. (22.2 mm)
Synonym(s):
Spinfin® magnetic stirring bar
All Photos(4)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
Recommended Products
material
PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37125-0078
H × diam.
0.5 in. (12.7 mm) × 0.875 in. (22.2 mm)
General description
Disks have fins on top for complete agitation.
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks, where other stirring bars cannot be used.
Features and Benefits
- Maintains its position in a vessel and will not wobble
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE® brand PTFE Coating – Superior Chemical Compatibility
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe
Legal Information
Spinfin is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company