BAF371090033
SP Bel-Art® Spinbar® magnetic stir bar
octagonal blue PTFE-coated, L × diam. 3 in. (76.2 mm) × 0.5 in. (12.7 mm)
Synonym(s):
Spinbar® magnetic stir bar
All Photos(8)
About This Item
Recommended Products
material
octagonal blue PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37109-0033
L × diam.
3 in. (76.2 mm) × 0.5 in. (12.7 mm)
Related Categories
General description
- Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
- Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
- Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
- Autoclave safe.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape. Choose from red, yellow or blue.
Features and Benefits
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE® brand PTFE Coating – Superior Chemical Compatibility
- Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
- Pivot ring aids in reducing friction and chattering
- Autoclavable
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)