Skip to Content
Merck
All Photos(4)

Documents

A26209

Sigma-Aldrich

Adenosine 5′-triphosphate disodium salt hydrate

99%

Synonym(s):

5′-ATP-Na2, ATP disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5Na2O13P3 · xH2O
CAS Number:
Molecular Weight:
551.14 (anhydrous basis)
Beilstein:
6264880
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

176 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[H]O[H].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O13P3.2Na.H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1

InChI key

NTBQNWBHIXNPRU-MSQVLRTGSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 5′-triphosphate disodium salt hydrate has been used as a component of mechanistic target of rapamycin (mTOR) assay start buffer. It has also been used in a feeding behavior study of Rhodnius prolixus as a potent phagostimulant in feeding solutions to enhance the feeding rate.

Other Notes

Contains solvent of crystallization, varying amounts of water

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro mTORC1 Kinase Assay for Mammalian Cells Protocol.
Dodd K M, et al.
Oncogene, 6 (2015)
Salt controls feeding decisions in a blood-sucking insect.
Pontes G, et al.
Journal of Insect Physiology, 98, 93-100 (2017)
Avishai Avni et al.
Frontiers in plant science, 11, 879-879 (2020-07-28)
Growth retardation and stress-induced premature plant senescence are accompanied by a severe yield reduction and raise a major agro-economic concern. To improve biomass and yield in agricultural crops under mild stress conditions, the survival must be changed to productivity mode.
András Szabó et al.
Toxins, 12(5) (2020-05-16)
Weaned piglets (n = 3 × 6) were fed 0, 15 and 30 mg/kg diet fumonisin (FB1, FB2 and FB3, i.e., FBs, a sphinganine analogue mycotoxin), from the age of 35 days for 21 days, to assess mycotoxin induced, dose-dependent
María J Rodríguez-López et al.
Insects, 11(9) (2020-09-13)
A combination of biological control and host plant resistance would be desirable for optimally controlling the greenhouse whitefly, Trialeurodes vaporariorum in tomato crops. Whitefly settlement preference, oviposition, and survivorship were evaluated on ABL 10-4 and 'Moneymaker', two nearly-isogenic tomato lines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service