Skip to Content
Merck
All Photos(1)

Documents

938661

Sigma-Aldrich

N-[2-(2-Hydroxyethoxy)ethyl]methacrylamide

new

contains MEHQ as inhibitor

Synonym(s):

N Methacrylamide, N-[2-(2-Hydroxyethoxy)ethyl]-2-methyl-2-propenamide, Cross linkers, Drug delivery crosslinker, HEEMAM, HEOEMA, Polyacetals, Stimuli-responsive, Trigger cleavable crosslinker, Water-soluble crosslinker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO3
CAS Number:
Molecular Weight:
173.21
UNSPSC Code:
12162008

form

liquid (or paste)

Quality Level

contains

MEHQ as inhibitor

color

yellow

storage temp.

−20°C

General description

N-[2-(2-Hydroxyethoxy)ethyl]methacrylamide can be used to synthesize polyacetal polymers. Tri(ethylene glycol) (N-hydroxyethoxy)ethyl methacrylamide which is a pH sensitive crosslinker and monomer containing acetal group can be synthesized from N-[2-(2-Hydroxyethoxy)ethyl]methacrylamide.

Application

  • Targeted drug delivery application
  • Water soluble and cross linkable
  • Stimuli-responsive hydrogels *
  • Radical polymerization

Features and Benefits

  • Water soluble and high water absorption
  • Biodegradable and biocompatible

related product

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Temperature-responsive polymeric sensors based on triphenylamine linked naphthalimide with red aggregation-induced emission
Zhou H, et al.
Dyes and Pigments, 216, 111356-111356 (2023)
Determination of Oxygen Permeability in Acrylic-Based Hydrogels by Proton NMR Spectroscopy and Imaging
Compa? V, et al.
Macromolecular Chemistry and Physics, 215, 624-637 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service