Skip to Content
Merck
All Photos(2)

Key Documents

938130

Sigma-Aldrich

Chloroplatinic acid hydrate

greener alternative

Technipur®, ≥99.9% trace metals basis

Synonym(s):

Hexachloroplatinic(IV) acid hydrate, Hydrogen hexachloroplatinate(IV) hydrate, Platinic chloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2PtCl6 · xH2O
CAS Number:
Molecular Weight:
409.81 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302

Quality Level

Assay

≥99.9% trace metals basis

form

powder, crystals or chunks

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤1000.0 ppm (trace metals analysis)

mp

60 °C (lit.)

density

2.43 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

O.Cl.Cl.Cl[Pt](Cl)(Cl)Cl

InChI

1S/6ClH.H2O.Pt/h6*1H;1H2;/q;;;;;;;+4/p-4

InChI key

SLIOYUPLNYLSSR-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Chloroplatinic acid hydrate is a popular platinum precursor which is used as catalyst for polymer electrolyte fuel cells, in sensors.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly sensitive, flexible H2 gas sensors based on less platinum bimetallic Ni?Pt nanocatalyst-functionalized carbon nanofibers
Nair K G, et al.
ACS applied electronic materials, 3(4), 1621-1633 (2021)
Electrochemical biosensor based on dewdrop-like platinum nanoparticles-decorated silver nanoflowers nanocomposites for H2O2 and glucose detection
Huang ,Z et al.
Journal of the Electrochemical Society, 166(13), B1138-B1138 (2019)
Polyol synthesis of reduced graphene oxide supported platinum electrocatalysts for fuel cells: Effect of Pt precursor, support oxidation level and pH
Hoe L P, et al.
International Journal of Hydrogen Energy, 43(35), 16998-17011 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service