Skip to Content
Merck
All Photos(1)

Key Documents

935417

Sigma-Aldrich

β-D-Glucopyranosyl azide

≥95%

Synonym(s):

2-Acetamido-2-deoxy-3,4,6-tri-O-acetyl-β-D-glucopyranosyl azide, 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide, Glucopyranosyl azide, 2-acetamido-2-deoxy-, 3,4,6-triacetate, β-D-, Glucopyranosyl azide, 2-acetamido-2-deoxy-, triacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N4O8
CAS Number:
Molecular Weight:
372.33
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder or crystals

color

white to light yellow

bp

501.81  °C

mp

128-130 °C

density

1.30 g/mL

storage temp.

−20°C

SMILES string

[N-]=[N+]=NC1OC(COC(=O)C)C(OC(=O)C)C(OC(=O)C)C1NC(=O)C

InChI

InChI=1S/C14H20N4O8/c1-6(19)16-11-13(25-9(4)22)12(24-8(3)21)10(5-23-7(2)20)26-14(11)17-18-15/h10-14H,5H2,1-4H3,(H,16,19)/t10-,11-,12-,13-,14-/m1/s1

InChI key

RMCFMPMNMQZHSF-DHGKCCLASA-N

Application

β-D-Glucopyranosyl azide, 2-(acetylamino)-2-deoxy-, 3,4,6-triacetate is commonly used in organic synthesis, particularly for the preparation of glycosyl azides, which are versatile intermediates in the synthesis of glycoconjugates. In carbohydrate chemistry, it can be employed for modifying and functionalizing carbohydrates, including the introduction of azide groups. These azide-modified carbohydrates can participate in click chemistry reactions, enabling the development of bioconjugates and glycoarrays. In drug discovery, this compound aids in the design and synthesis of carbohydrate-based therapeutics, such as glycosidase inhibitors or glycoconjugate vaccines, by modifying carbohydrate structures. For biochemical research, this compound is valuable in studying carbohydrate-protein interactions, glycan biosynthesis, and glycoprotein engineering. It allows for the modification and labeling of carbohydrates for subsequent biological assays and analyses. Finally, as an azide derivative, it finds application in click chemistry reactions, specifically the azide-alkyne cycloaddition reaction (commonly known as the "click reaction"). This reaction enables efficient and selective labeling, bioconjugation, and cross-linking in various biological and materials science applications.

Features and Benefits

β-D-Glucopyranosyl azide, 2-(acetylamino)-2-deoxy-, 3,4,6-triacetate is a complex carbohydrate and the glycosylation product of 2,3,4,6-tetraacetyl α--D--glucose and 2,3,6 -tri--O--acetyl--2--deoxy--β--D--glucopyranose. This compound has been modified by Click Chemistry with 4-(dimethylamino)pyridine (DMAP). The modification has produced an acetamido group at the C2 position of the glucopyranoside moiety. The compound is available in high purity for research purposes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Biological Evaluation of Novel Carbohydrate-Derived Derivatives of Erlotinib
Yu, Wenbo
Drug Development Research, 77, 319-325 (2016)
Georg C Rudolf et al.
Chembiochem : a European journal of chemical biology, 14(18), 2447-2455 (2013-10-30)
Copper-catalysed alkyne-azide 1,3-dipolar cycloaddition (CuAAC) is the predominantly used bioconjugation method in the field of activity-based protein profiling (ABPP). Several limitations, however, including conversion efficiency, protein denaturation and buffer compatibility, restrict the scope of established procedures. We introduce an ABPP
Wenbo Yu et al.
Drug development research, 77(6), 319-325 (2016-08-16)
Preclinical Research A series of novel carbohydrate-derived Erlotinib derivatives were prepared by the copper-catalyzed cycloaddition reaction of erlotinib with various azido-sugars. The structures of the newly synthesized compounds were characterized and their cytostatic effects evaluated in vitro on human cancer
Brendan L Wilkinson et al.
Journal of medicinal chemistry, 51(6), 1945-1953 (2008-03-01)
A library of glycoconjugate benzene sulfonamides have been synthesized and investigated for their ability to inhibit the enzymatic activity of physiologically relevant human carbonic anhydrase (hCA) isozymes: hCA I, II, and tumor-associated IX. Our synthetic strategy directly links the known
Inhibition of carbonic anhydrases with glycosyltriazole benzene sulfonamides
Wilkinson, Brendan
Journal of Medicinal Chemistry, 51, 1945-1953 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service