Skip to Content
Merck
All Photos(1)

Key Documents

935018

Sigma-Aldrich

Carbon Fiber Microelectrode

diam. 11 μm , (8 – 13 µm)

Synonym(s):

MF-2007

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NA.21
Pricing and availability is not currently available.

diam.

11 μm , (8 – 13 µm)

General description

Microelectrodes consist of small diameter metal wires or fibers (5-100 µm) sealed within tempered glass bodies.
We offer 11 µm (+/- 2 um) diameter Carbon Fiber Microelectrode (Working electrodes) for R&D applications in electrochemistry.
Dimensions of the electrode are approximately 7 cm long and 4 mm OD. The electrical connection is a 0.060" gold-plated pin. Each microelectrode is visually inspected and electrochemically tested before shipping.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service