Skip to Content
Merck
All Photos(1)

Key Documents

93412

Sigma-Aldrich

Tris(trimethylsilyl) phosphite

≥95.0% (GC)

Synonym(s):

Trimethylsilanol phosphite, Tris(trimethylsilyloxy)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3SiO]3P
CAS Number:
Molecular Weight:
298.54
Beilstein:
2043814
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.409 (lit.)

bp

78-81 °C/8 mmHg (lit.)

density

0.893 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C

InChI

1S/C9H27O3PSi3/c1-14(2,3)10-13(11-15(4,5)6)12-16(7,8)9/h1-9H3

InChI key

VMZOBROUFBEGAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(trimethylsilyl)phosphate is tris(trimethylsilyl)ester of phosphoric acid. It is an important raw material and intermediate inorganic synthesis. It is used in the preparation of bis(trimethylsilyl)phosphonate esters by the Arbuzov reaction with halogen-containing compounds.

Application

Tris(trimethylsilyl) phosphite is used as a:
  • Film-forming additive to enhance the electrochemical performance of the NMC811[LiNi0.8Mn0.1Co0.1O2]/Si-Grcell.

Other Notes

Reagent used for the mild preparation of bis(TMS) phosphonates (Arbuzow reaction); Review

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Understanding the Roles of Tris (trimethylsilyl) Phosphite (TMSPi) in LiNi0. 8Mn0. 1Co0. 1O2 (NMC811)/Silicon--Graphite (Si--Gr) Lithium-Ion Batteries
Liu H, et al.
Advanced Materials Interfaces, 7(15), 2000277-2000277 (2020)
Tris (trimethylsilyl) Phosphite
Bittman R
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
L.E. Dreef et al.
Tetrahedron Letters, 29, 1199-1199 (1988)
M. Sekine et al.
The Journal of Organic Chemistry, 46, 2097-2097 (1981)
M. Sekine et al.
Journal of the Chemical Society. Chemical Communications, 285-285 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service