Skip to Content
Merck
All Photos(1)

Key Documents

921750

Sigma-Aldrich

endo-N-Methyl-5-norbornene-2-carboxamide

≥95%

Synonym(s):

endo-N-Methylbicyclo[2.2.1]hept-5-ene-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

mp

105-108 °C

storage temp.

2-8°C

Application

endo-N-Methyl-5-norbornene-2-carboxamide is a norbornene ligand developed in the lab of Guanbing Dong for use in norbornene/palladium catalysis.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhe Dong et al.
Angewandte Chemie (International ed. in English), 54(43), 12664-12668 (2015-09-04)
Reported herein is a palladium/norbornene-catalyzed ortho-arene acylation of aryl iodides by a Catellani-type C-H functionalization. This transformation is enabled by isopropyl carbonate anhydrides, which serve as both an acyl cation equivalent and a hydride source.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service