Skip to Content
Merck
All Photos(1)

Key Documents

906921

Sigma-Aldrich

PQT-12

Synonym(s):

Poly(3,3′′′-didodecyl[2,2′:5′,2′′:5′′,2′′′-quaterthiophene]-5,5′′′-diyl), Poly(4,4′′-didodecyl[2,2′:5′,2′′:5′′,2′′′-quaterthiophene]-5,5′′′-diyl)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C40H56S4)n
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

Band gap: 2.27 eV
Eox = 0.45 V
Solution processable (chloroform, chlorobenzene, etc)
LIFT transferable

form

solid

mol wt

Mw 10,000-25,000 by GPC

color

brown

solubility

>5 mg/mL (in CHCl3)

λmax

473 nm in toluene

Orbital energy

HOMO -5.24 eV 
LUMO -2.97 eV 

storage temp.

15-25°C

Application

PQT-12 can be employed as the semiconductor material in organic field-effect transistors(OFETs). Its high charge carrier mobility, good film-forming properties, and stability make it useful for the channel layer, enabling efficient electronic device performance. The conjugated polymer structure and charge storage capability of PQT-12 make it potentially useful for energy storage applications such as super capacitors and batteries.
PQT-12 exhibits strong absorption in the visible and near-infrared regions of the spectrum. This property enables effective utilization of a broad range of solar radiation, enhancing the light-harvesting capability and efficiency of OPV devices. PQT-12 can serve as the donor material in the active layer of OPV devices. PQT-12′s unique properties and charge transport characteristics contribute to the development of low voltage and low-power OFET devices. Its high charge carrier mobility allows for better device performance with lower operational voltages.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of the Poly(bisdodecylquaterthiophene) Structure for High and Predominantly Nonionic Conductivity with Matched Dopants
Hui Li, et al.
Journal of the American Chemical Society, 139,, 11149- 11157 (2017)
20 megahertz operation of organic nanodiodes.
Majewski L A, et al.
Physica Status Solidi B, 253(8), 1507-1510 (2016)
Highly Flexible Organic Nanofiber Phototransistors Fabricated on a Textile Composite for Wearable Photosensors
Lee M Y, et al.
Advances in Functional Materials, (2016), 1445-1445 (2016)
Highly flexible chemical sensors based on polymer nanofiber field-?effect transistors
Kweon O Y, et al
Journal of Material Chemistry C (2019)
Laser printed organic semiconductor PQT-12 for bottom-gate organic thin-film transistors: Fabrication and characterization
Makrygianni M, et al.
Journal of Chemical and Pharmaceutical Sciences , 390, 823-830 (2016)

Articles

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service