Recommended Products
form
solid
General description
Tungsten (IV) diselenide is a p-type 2D semiconducting transition metal dichalcogenide (TMD) with an energy bandgap of 1-2 eV. It can be used in the fabrication of optoelectronic devices.
Application
Tungsten (IV) diselenide can be used for a variety of applications such as fiber laser, heterojunction solar cells, and lithium batteries.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tungsten diselenide Q-switched erbium-doped fiber laser
Optical Engineering, 55(8), 081306-081306 (2016)
A systematic study of the synthesis of monolayer tungsten diselenide films on gold foil
Current Applied Physics, 16(9), 1216-1222 (2016)
Liquid-solid surface phase transformation of fluorinated fullerene on monolayer tungsten diselenide
Nanotechnology, 29(17), 174002-174002 (2018)
Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets
The Journal of Physical Chemistry C, 118 (8), 3847-3863 (2014)
Potential application of multilayer n-type tungsten diselenide (WSe2) sheet as transparent conducting electrode in silicon heterojunction solar cell
Computational Materials Science, 136(17), 102-108 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service