Skip to Content
Merck
All Photos(1)

Key Documents

903841

Sigma-Aldrich

Tungsten (IV) disulfide

lithium intercalated

Synonym(s):

LiWS2, Lithium intercalated 2D-Tungsten disulfide, Lithium intercalated tungsten sulfide, WS2Li

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LixWS2
CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

Application

Tungsten (IV) disulfide is a transition metal dichalcogenide that is a layered semiconductor that can be used as an anode material for lithium ion batteries by atomic layered deposition (ALD). It can also be used in potential applications in photovoltaic solar cells (PSCs) or lubricants.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of tungsten disulfide (WS2) nanoflakes for lithium ion battery application
Feng C, et al.
Electrochemical Communications, 9(1), 119-122 (2007)
Intercalation based tungsten disulfide (WS 2) Li-ion battery anode grown by atomic layer deposition
Nandi DK, et al.
Royal Society of Chemistry Advances, 6(44), 38024-38032 (2016)
Characterisation of rf sputtered tungsten disulfide and oxysulfide thin films
Martin-Litas I, et al.
Thin Solid Films, 416(1-2), 1-9 (2002)
Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
J L Gunjakar et al.
The Journal of Physical Chemistry C, 118, 3847-3847 (2014)
Tungsten disulfide nanotubes for lithium storage
Wang GX, et al.
Electrochemical and Solid-State Letters, 7(10), A321-A323 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service