Skip to Content
Merck
All Photos(1)

Key Documents

902667

Sigma-Aldrich

Propargyl-PEG6-acid

Synonym(s):

4,7,10,13,16,19-Hexaoxadocos-21-ynoic acid, Propargyl-PEG6-CH2CO2H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28O8
CAS Number:
Molecular Weight:
348.39
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Assay

≥95%

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

refractive index

n/D 1.469

functional group

alkyne
carboxylic acid

storage temp.

−20°C

SMILES string

OC(CCOCCOCCOCCOCCOCCOCC#C)=O

InChI

1S/C16H28O8/c1-2-4-19-6-8-21-10-12-23-14-15-24-13-11-22-9-7-20-5-3-16(17)18/h1H,3-15H2,(H,17,18)

InChI key

XRSJYQQOJNYUDV-UHFFFAOYSA-N

Application

This heterobifunctional, PEGylated crosslinker features a carboxylic acid at one end and propargyl group at the other for reaction with azide-containing compounds using click chemistry. The hydrophilic PEG linker facilitates solubility in biological applications. Propargyl-PEG6-acid can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.

Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service