Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
901823
Nickel electroplating solution
Synonym(s):
Ni electroplating solution, Ni plating solution
About This Item
Recommended Products
form
liquid
contains
1.25 g/L±0.125 g/L stabilizer
0.015 g/L±0.0015 g/L wetting agent
4.25 g/L±0.25 g/L Stress reducer
composition
30 g/L ±0.5 g/L H3BO3
170g/L ±5 g/L Ni(SO3NH2)2.4H2O
9.51 g/L ±1 g/L NiBr2 .xH2O
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Deposits have low internal stress and are very ductile.
- Surfaces are uniform and smooth with well-controlled thickness and excellent uniformity.
- As plated Ni films have small grains in the range of 10 - 50 nm. Grain size orientations are (111) and (200) for FCC (face centred cubic).
- Qualified bath metrology for replenishment and a long shelf life.
Electroplated Ni film (5-10 μm thickness) properties:
- Density: 8.1 g/cm3
- Young′s modulus: 210 GPa
- Internal stress 33 ± 13 MPa
- Tensile strength: 585 ± 40 MPa
- Grain size: 10-50 nm
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Carc. 1A Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Inhalation
Target Organs
Respiratory Tract
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service