Skip to Content
Merck
All Photos(1)

Documents

900221

Sigma-Aldrich

CdSe/ZnS core-shell type quantum dots

carboxylic acid functionalized, fluorescence λem 540 nm, 1 mg/mL in H2O

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSe/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

form

powder

Quality Level

concentration

1 mg/mL in H2O

fluorescence

λem 540 nm
λem 540 nm±10 nm, quantum yield >50%

functional group

carboxylic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Reactive group: Carboxylic acid.

Application

The organic layer of the CdS/ZnS or CdSe/ZnS quantum dots (QDs) with carboxylic acid as reactive group consist of a monolayer of oleic acid/octadecylamine and a monolayer of amphiphilic polymer. The total thickness of organic layers on these QDs is approximately 4 nm. They find many applications in bioimaging and biosensing.

Caution

Do not freeze.

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingjiang Sun et al.
ACS nano, 3(3), 737-743 (2009-03-25)
Owing to their narrow bright emission band, broad size-tunable emission wavelength, superior photostability, and excellent flexible-substrate compatibility, light-emitting diodes based on quantum dots (QD-LEDs) are currently under intensive research and development for multiple consumer applications including flat-panel displays and flat
Zhanao Tan et al.
Nano letters, 7(12), 3803-3807 (2007-11-03)
We report a multilayer solution-processed blue light-emitting diode based on colloidal core/shell CdS/ZnS nanocrystal quantum dots (QDs). At a low-operating voltage of 5.5 V, the device emits spectrally pure blue radiation at 460 nm with a narrow full-width-at-half-maximum bandwidth of
Bright, multicoloured light-emitting diodes based on quantum dots.
Sun Q, et al.
Nature Photonics, 1, 717-722 (2007)

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service