Skip to Content
Merck
All Photos(1)

Key Documents

87303

Sigma-Aldrich

Tetrahexylammonium hydrogensulfate

≥98.0% (T)

Synonym(s):

THAHS, Tetra-n-Hexylammonium Hydrogen Sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)5]4N(HSO4)
CAS Number:
Molecular Weight:
451.75
Beilstein:
4629729
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (T)

form

crystals

mp

98-100 °C (lit.)

SMILES string

OS([O-])(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

InChI

1S/C24H52N.H2O4S/c1-5-9-13-17-21-25(22-18-14-10-6-2,23-19-15-11-7-3)24-20-16-12-8-4;1-5(2,3)4/h5-24H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

RULHPTADXJPDSN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tetrahexylammonium hydrogensulfate can be used as a phase-transfer agent to facilitate certain organic reactions in a two-liquid phase system. For example:
  • Epoxidation of α,β-unsaturated ketones in a biphasic dichloromethane/water mixture using sodium perborate.
  • Transfer hydrogenolysis of aryl bromides by aqueous sodium formate catalyzed by palladium-triphenylphospine.
  • Carbonylation of benzylic and allylic bromides to synthesize carboxylic esters.

Other Notes

Lipophilic phase transfer catalyst. Most efficient catalyst in ruthenium-catalyzed reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Bar et al.
Tetrahedron Letters, 22, 1709-1709 (1981)
Transfer hydrogenolysis of aryl bromides by sodium formate and palladium?phosphine catalyst under phase transfer conditions.
Bar R, et al.
J. Mol. Catal., 16(2), 175-180 (1982)
Epoxidation of ?, ?-unsaturated ketones with sodium perborate.
Straub T S, et al.
Tetrahedron Letters, 36(5), 663-664 (1995)
Catalytic carbonylation of benzylic and allylic bromides by a rhodium zwitterionic complex under phase transfer catalysis conditions
Amaratunga S and Alper H
Journal of Organometallic Chemistry, 488, 25-28 (1995)
K M Sunasara et al.
Biotechnology progress, 17(5), 897-906 (2001-10-06)
This work investigates the utility of RPLC displacement chromatography for the purification of recombinant brain derived neurotrophic factor (rHu-BDNF) from its variants and E. coli. protein (ECP) impurities. The closely associated variants (six in total) differ by one amino acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service