Skip to Content
Merck
All Photos(1)

Key Documents

798746

Sigma-Aldrich

Exo-4-anisole Kwon [2.2.1] bicyclic phosphine

Synonym(s):

Exo-4-Methoxyphenyl Kwon [2.2.1] Bicyclic Phosphine, (1S,4S,5R)-5-(4-methoxyphenyl)-2-tosyl-2-aza-5-phosphabicyclo[2.2.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22NO3PS
Molecular Weight:
375.42
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

mp

85-90 °C

functional group

phosphine
sulfonamide

SMILES string

O=S(N1C[C@@]2([H])[P@](C3=CC=C(OC)C=C3)C[C@]1([H])C2)(C4=CC=C(C)C=C4)=O

InChI

1S/C19H22NO3PS/c1-14-3-9-19(10-4-14)25(21,22)20-12-18-11-15(20)13-24(18)17-7-5-16(23-2)6-8-17/h3-10,15,18H,11-13H2,1-2H3/t15-,18-,24?/m0/s1

InChI key

RMFZMNUGIFSNIE-YPAHSFBSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The bicyclic, chiral phosphine was developed by the Kwon Research Group.Its initial applications were for asymmetric [3+2] annulation between allenes and imines as well as the first examples of phosphine-catalyzed asymmetric syntheses of 1,2,3,5-substituted pyrrolines. Along with nucleophilic organocatalysis, the P-chiral phosphines may also find utility in asymmetric transition-metal catalysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service