771368
2-Methyl-2-cyclohexen-1-one
≥90%
Synonym(s):
2-Methyl-2-cyclohexene-1-one, 2-Methyl-2-cyclohexenone, 2-Methyl-3-oxocyclohexene
About This Item
Assay
≥90%
form
liquid
refractive index
n20/D 1.487
bp
176-179 °C/760 mmHg
density
0.972 g/mL at 25 °C
functional group
ketone
SMILES string
CC1=CCCCC1=O
InChI
1S/C7H10O/c1-6-4-2-3-5-7(6)8/h4H,2-3,5H2,1H3
InChI key
LKTNAAYQZJAXCJ-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
140.0 °F
Flash Point(C)
60 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service