769916
Resomer® LR 704 S, Poly(L-lactide-co-D,L-lactide)
ester terminated, L-Lactide: D,L-Lactide 70:30
Synonym(s):
Resomer® LR 704 S, P(L/DL)LA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
solid
feed ratio
lactide:lactide 70:30
viscosity
2.0-2.8 dL/g
mp
102-111 °C
transition temp
Tg 56-62 °C
storage temp.
2-8°C
Application
Biodegradable, biocompatible, and bioresorbable copolymer composed of L-Lactide and D,L-Lactide. This semi-crystalline material has been used in the fabrication of research medical devices and research tissue engineering solutions, such as orthopedic or soft tissue fixation devices. Degradation of this materials has been thoroughly studied and has been shown to be safely resorbed by the body after implantation. Modification of molecular weight and polymer composition allows for control of the degradation rate and mechanical stability of the polymer.
Legal Information
RESOMER is a registered trademark of Evonik Rohm GmbH
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service