Skip to Content
Merck
All Photos(3)

Key Documents

768618

Sigma-Aldrich

Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate)

dry re-dispersible pellets

Synonym(s):

Orgacon DRY, PEDOT:PSS, Poly(2,3-dihydrothieno-1,4-dioxin)-poly(styrenesulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

dry re-dispersible pellets

resistance

200-450 Ω/sq

mp

>300 °C

Looking for similar products? Visit Product Comparison Guide

General description

A conducting polymer such as poly(3,4-ethylenedioxythiophene) doped with poly(styrene sulfonate) anions (PEDOT/PSS) is widely used in various organic optoelectronic devices. PEDOT: PSS is a blend of cationic polythiopene derivative, doped with a polyanion. High electrical conductivity and good oxidation resistance of such polymers make it suitable for electromagnetic shielding and noise suppression. Thus, the polymer film was found to possess high transparency throughout the visible light spectrum and even into near IR and near UV regions, virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Impact of small electric and magnetic fields on the polymer was studied.
Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate) (PEDOT:PSS) is a conductive polymer (CP) that is formed by electropolymerizing 3,4-ethylenedioxythiophene in a solution of poly(styrenesulfonate) (PSS). PEDOT is doped with positive ions and PSS with negative ions. It is a low cost CP that is produced with physical appearance (for example dispersible pellets). The following are the properties that make PEDOT:PSS a viable polymer in organic electronics:
  • good optical properties
  • high conductivity
  • functionalization of nanomaterials
  • high thermal stability
  • easy processing

Application

Orgacon DRY is specifically useful for water sensitive formulations containing UV curable monomer (suitable with small amount of water) and pre-polymer based formulations.

Orgacon DRY pellets can be re-dispersed in organic solvents such as ethanol. The resulting dispersion can be used in coating - or ink - formulations targeted for different applications.

Allow retention of PEDOT/PSS key properties in polar organic solvents; such as Ethanol; Surface Resistance: 200~450 Ohm/square; Visual Light Transmission: ~85%
PEDOT:PSS can be used as a conductive material with a high mobility for charge carriers. It can be used for a wide range of energy based applications such as organic printable electronics, textile electronics, electrochemical storage and supercapacitors, and lithium ion batteries.
Virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Conductive polymer blend.

Physical properties

  • Active chemical PEDOT/PSS
  • Visual appearance blue-grey pellets
  • Water content max. 15 % under 50% RH; 22°C
  • Shelf life 24 month in storage conditions

Preparation Note

When spin-coated using S300 EtOH (contains less than 3.5% wt. H20) as the solvent; the Visual Light Transmission (VLT) according to ASTM D 1003 (excludes substrate) is 85%; and the Surface Resistance is 450 Ohm/square.

Legal Information

Product of Agfa-Gevaert N.V.
Orgacon is a trademark of Agfa-Gevaert N.V.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Foldable Textile Electronic Devices Using All-Organic Conductive Fibers.
Miura H, et al.
Advanced Engineering Materials, 16(5), 550-555 (2014)
Wet-spinning of PEDOT: PSS/functionalized-SWNTs composite: a facile route toward production of strong and highly conducting multifunctional fibers.
Jalili R, et al.
Scientific reports, 3(18), 3438-3438 (2013)
Exploiting high quality PEDOT: PSS-SWNT composite formulations for wet-spinning multifunctional fibers.
Jalili R, et al.
Journal of Materials Chemistry, 22(48), 25174-25182 (2012)
Synthesis and optimization of PEDOT: PSS based ink for printing nanoarrays using Dip-Pen Nanolithography.
Wagner M, et al.
Synthetic Metals, 181(18), 64-71 (2013)
Three dimensional cellular architecture of sulfur doped graphene: self-standing electrode for flexible supercapacitors, lithium ion and sodium ion batteries.
Islam MM, et al.
Journal of Material Chemistry A, 5(11), 5290-5302 (2017)

Articles

A detailed article on conducting polymer materials for flexible organic photovoltaics (OPVs) applications.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service