Skip to Content
Merck
All Photos(2)

Key Documents

765198

Sigma-Aldrich

Lithium manganese nickel oxide

greener alternative

electrode sheet, aluminum substrate, size 5 in. × 10 in.

Synonym(s):

LMNO, Lithium nickel manganate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2Mn3NiO8
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

description

Nominal Voltage: 4.7 V, Li/Li+

Assay

≥98%

form

solid

composition

loading, ≥80%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

≥80% loading

size

5 in. × 10 in.

thickness

25-50 μm

particle size

4-7 μm (typical)

capacity

115 mAh/g(minimum)
125 mAh/g(nominal at 0.1C)

mp

>1000 °C

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[Li+].[O-][Mn]=O.[O-][Ni]=O.O=[Mn]=O.O=[Mn]=O

InChI

1S/2Li.3Mn.Ni.8O/q2*+1;;;;;;;;;;;2*-1

InChI key

NTWFBJKNXFUJHM-UHFFFAOYSA-N

General description

Lithium manganese nickel oxide (LMNO) is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

LMNO is a spinel cathode material that has a storage capacity greater than 200 mAhg−1 at a voltage around 4.7 V. It has a high irreversible capacity due to its inherent property to self-discharge. LMNO can be used in the fabrication of high-performance lithium-ion batteries.
LMNO is cathode material with good electrochemical performance for high power and high energy density Lithium ion batteries. LMNO can can intercalate Li reversibly at a high voltages.

The LMNO casted electrode sheets can be cut into appropriate size and is ready to be used in lithium ion batteries.

Other Notes

Crystal Structure: Cubic

Operating Condiditons:
  • Recommended maximum charge voltage: 5.0 V vs Li/Li+
  • Recommended maximum charge current: 5 C
  • Recommended cut-off voltage for discharge: 3.5 V vs Li/Li+
  • Recommended maximum discharge current: 5 C
  • Recommended charge method: constant current - constant voltage

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Liu, D.; et al.
Journal of Power Sources, 204, 127-127 (2012)
High capacity lithium-manganese-nickel-oxide composite cathodes with low irreversible capacity loss and good cycle life for lithium ion batteries
Zhang J, et al.
Science China: Chemistry, 59(11), 1479-1485 (2016)
Surface chemistry of metal oxide coated lithium manganese nickel oxide thin film cathodes studied by XPS
Baggetto L, et al.
Electrochimica Acta, 90, 135-147 (2013)
Lithium-manganese-nickel-oxide electrodes with integrated layered-spinel structures for lithium batteries
Park S-H, et al.
Electrochemical Communications, 9(2), 262-268 (2007)
Markovsky, B.; et al.
Electrochemical Communications, 6, 821-821 (2004)

Articles

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service