Skip to Content
Merck
All Photos(1)

Key Documents

761524

Sigma-Aldrich

Dibenzocyclooctyne-N-hydroxysuccinimidyl ester

for Copper-free Click Chemistry

Synonym(s):

DBCO-NHS ester, DBCO-SE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H18N2O5
Molecular Weight:
402.40
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

mp

149-157 (decomposition)

functional group

NHS ester

storage temp.

−20°C

SMILES string

O=C(CCC(ON(C(CC1)=O)C1=O)=O)N2CC3=C(C=CC=C3)C#CC4=C2C=CC=C4

InChI

1S/C23H18N2O5/c26-20(13-14-23(29)30-25-21(27)11-12-22(25)28)24-15-18-7-2-1-5-16(18)9-10-17-6-3-4-8-19(17)24/h1-8H,11-15H2

InChI key

XCEBOJWFQSQZKR-UHFFFAOYSA-N

General description

DBCO-NHS ester is commonly used in bioconjugation. It consists of an NHS ester that reacts with primary amines on biomolecules and an alkyne group that allows for a specific reaction with molecules containing azides. This reaction is carried out through a copper-free click chemistry process called strain-promoted alkyne-azide cycloaddition.

Application

Dibenzocyclooctyne-N-hydroxysuccinimidyl ester is commonly used:
  • In the modification and labeling of biomolecules such as antibodies and streptavidin by introducing DBCO groups onto their surfaces
  • As a cross linker to conjugate peptide antigens onto the surface of poly(lactic-co-glycolic acid) (PLGA) nanoparticles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Ma et al.
Biomaterials, 255, 120205-120205 (2020-06-24)
Three biorthogonal click reactions, a photoinitiated thiol-yne reaction, an azide-alkyne cycloaddition, and a methyltetrazine-transcyclooctene Diels Alder, were used to independently control the presentation of several bioactive proteins to valvular interstitial cells (VICs) in hydrogel scaffolds. Tethered fibroblast growth factor (FGF-2)
Nanoparticles functionalized with collagenase exhibit improved tumor accumulation in a murine xenograft model.
Murty S, et al.
Particle & Particle Systems Characterization, 31(12), 1307-1312 (2014)
Preparation of single-and double-oligonucleotide antibody conjugates and their application for protein analytics
J Wiener, et al.
Scientific reports, 10, 1457-1457 (2020)
Investigating functional DNA grafted on nanodiamond surface using site-directed spin labeling and electron paramagnetic resonance spectroscopy.
Akiel RD., et al.
The Journal of Physical Chemistry B, 120(17), 4003-4008 (2016)
Graphene Oxide Nanosheets Modified with Single-Domain Antibodies for Rapid and Efficient Capture of Cells.
Chen GY, et al.
Chemistry?A European Journal , 21(48), 17178-17183 (2015)

Articles

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service