Skip to Content
Merck
All Photos(1)

Documents

760161

Sigma-Aldrich

Guanosine-13C10,15N5 3′,5′-cyclic monophosphate calcium salt

99 atom % 13C, 98 atom % 15N, 95% (CP)

Synonym(s):

GMP-13C10,15N5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C10H1115N5O7P · .5Ca
Molecular Weight:
379.13
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

95% (CP)

form

solid

technique(s)

bio NMR: suitable

mass shift

M+15

storage temp.

−20°C

SMILES string

[15NH2][13C]([15NH]1)=[15N][13C]2=[13C]([15N]=[13CH][15N]2[13C@H]3[13C@H](O)[13CH](O4)[13C@@H]([13CH2]O[P@@]4([O-])=O)O3)[13C]1=O.[15NH2][13C]([15NH]5)=[15N][13C]6=[13C]([15N]=[13CH][15N]6[13C@H]7[13C@H](O)[13CH](O8)[13C@@H]([13CH2]O[P@@]8([O-])=O)O7)[13C

InChI

1S/2C10H12N5O7P.Ca/c2*11-10-13-7-4(8(17)14-10)12-2-15(7)9-5(16)6-3(21-9)1-20-23(18,19)22-6;/h2*2-3,5-6,9,16H,1H2,(H,18,19)(H3,11,13,14,17);/q;;+2/p-2/t2*3-,5-,6-,9-;/m11./s1/i2*1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1;

InChI key

UWPRAAHTBFUSLT-NDZKCIGYSA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service