Skip to Content
Merck
All Photos(2)

Key Documents

759597

Sigma-Aldrich

4,4′-Stilbenedicarboxylic acid

greener alternative

98%

Synonym(s):

4,4′-(1,2-Ethenediyl)bisbenzoic acid, 4,4′-Dicarboxystilbene, Biphenylethene-4,4·-dicarboxylic acid, H2BPEA, H2SDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O4
CAS Number:
Molecular Weight:
268.26
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

400-420 °C

greener alternative category

SMILES string

OC(=O)c1ccc(\C=C\c2ccc(cc2)C(O)=O)cc1

InChI

1S/C16H12O4/c17-15(18)13-7-3-11(4-8-13)1-2-12-5-9-14(10-6-12)16(19)20/h1-10H,(H,17,18)(H,19,20)/b2-1+

InChI key

SBBQDUFLZGOASY-OWOJBTEDSA-N

General description

4,4′-Stilbenedicarboxylic acid (SDA) is a luminescent ligand that is useful as an organic linker due to the presence of two carboxylic groups, which form bridges between metallic centers. It has conjugated aromatic rings that facilitate the formation of a rigid linker.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

SDA can be used in the hydrothermal synthesis of three-dimensional (3D) metal-organic frameworks, which have potential usage in sensing, drug delivery, luminescence, and gas adsorption. It may also be used in energy storage applications such as supercapacitors and batteries.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chengfeng Yu et al.
Dalton transactions (Cambridge, England : 2003), 49(34), 12082-12087 (2020-08-22)
White light-emitting diodes (WLEDs) have aroused great attention due to their potential technological applications. In this work, we present two new Zn(ii) anthracene-linker-driven coordination polymers that exhibit intrinsic white-light emission. The emission covers the whole visible spectrum at room temperature.
An unprecedented stable self-penetrating metal-organic framework with solvent-dependent luminescence properties
Xiao Y, et al.
Inorganic Chemistry Communications, 59(5), 32-35 (2015)
Yadagiri Rachuri et al.
Inorganic chemistry, 58(17), 11389-11403 (2019-08-23)
We synthesized two new adenine-based Zn(II)/Cd(II) metal-organic frameworks (MOFs), namely, [Zn2(H2O)(stdb)2(5H-Ade)(9H-Ade)2] n
Marcel Schulz et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(59), 13598-13608 (2019-08-10)
In a systematic investigation, the synthesis of metal-organic frameworks (MOFs) with MIL-140 structure was studied. The precursors of this family of MOFs are the same as for the formation of the well-known UiO-type MOFs although the synthesis temperature for MIL-140
A series of three-dimensional lanthanide metal-organic frameworks with biphenylethene-4, 4?-dicarboxylic acid: Hydrothermal syntheses and structures
Deng Z, et al.
CrystEngComm, 12(5), 1526-1535 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service