Skip to Content
Merck
All Photos(2)

Key Documents

759287

Sigma-Aldrich

4-(Trifluoromethyl)cyclohexanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9F3O
CAS Number:
Molecular Weight:
166.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.400

density

1.221 at 25 °C

SMILES string

FC(F)(F)C1CCC(=O)CC1

InChI

1S/C7H9F3O/c8-7(9,10)5-1-3-6(11)4-2-5/h5H,1-4H2

InChI key

IPQDZFUZVJKAKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinglei Meng et al.
Nature communications, 8, 14190-14190 (2017-02-01)
Cyclohexanone and its derivatives are very important chemicals, which are currently produced mainly by oxidation of cyclohexane or alkylcyclohexane, hydrogenation of phenols, and alkylation of cyclohexanone. Here we report that bromide salt-modified Pd/C in H

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service