Skip to Content
Merck
All Photos(1)

Key Documents

75570

Sigma-Aldrich

Triethyl orthobenzoate

≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(OC2H5)3
CAS Number:
Molecular Weight:
224.30
Beilstein:
645341
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.472 (lit.)
n20/D 1.472

bp

239-241 °C (lit.)
80 °C/0.1 mmHg (lit.)

density

0.991 g/mL at 25 °C (lit.)

functional group

ether
phenyl

SMILES string

CCOC(OCC)(OCC)c1ccccc1

InChI

1S/C13H20O3/c1-4-14-13(15-5-2,16-6-3)12-10-8-7-9-11-12/h7-11H,4-6H2,1-3H3

InChI key

BQFPCTXLBRVFJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service