Skip to Content
Merck
All Photos(4)

Key Documents

753246

Sigma-Aldrich

RuPhos Pd G2

Synonym(s):

2nd Generation RuPhos Precatalyst, Chloro(2-dicyclohexylphosphino-2′,6′-diisopropoxy-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II), RuPhos-Pd-G2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H53ClNO2PPd
CAS Number:
Molecular Weight:
776.72
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

feature

generation 2

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

195-197 °C

functional group

phosphine

SMILES string

Nc1ccccc1-c2ccccc2[Pd]Cl.CC(C)Oc3cccc(OC(C)C)c3-c4ccccc4P(C5CCCCC5)C6CCCCC6

InChI

1S/C30H43O2P.C12H10N.ClH.Pd/c1-22(2)31-27-19-13-20-28(32-23(3)4)30(27)26-18-11-12-21-29(26)33(24-14-7-5-8-15-24)25-16-9-6-10-17-25;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h11-13,18-25H,5-10,14-17H2,1-4H3;1-6,8-9H,13H2;1H;/q;;;+1/p-1

InChI key

VPDRBFBKNSFRSS-UHFFFAOYSA-M

General description

RuPhos Pd G2 is a second generation (G2) precatalyst containing a biphenyl-based ligand. Product participates in various palladium catalyzed cross-coupling reactions, C-C, C-N, and C-O bond formation reactions and Suzuki-Miyaura coupling reactions. It generates active Pd catalyst at room temperature in the presence of weak phosphate or carbonate bases.

Application

RuPhos Pd G2 may be employed as precatalyst in the preparation of fluoro-substituted diazatetracenes and diazapentacenes, via Pd-catalyzed aryl amination reaction.

Other Notes

May contain <1% acetone.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonas Schwaben et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(39), 13758-13771 (2015-08-08)
Non-symmetrical 6,13-disubstituted pentacenes bearing trifluoromethyl and aryl substituents have been synthesized starting from pentacenequinone. Diazapentacenes with a variety of fluorine substituents were prepared either via a Hartwig-Buchwald aryl amination route or by a SNAr strategy. As a result of a

Articles

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

See All

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service